Sodium borohydride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
M-Clarity™ quality level = MQ200
632287-100G
632287-500G
SIAL632287-100GEA
121
EUR
SIAL632287-100G
SIAL632287-500G
Sodium borohydride, Sigma-Aldrich®
Sodium borohydride
Formula:
NaBH₄ MW: 37.83 g/mol Melting Pt: 400 Density: 1.07 g/cm³ (20 °C) Flash Pt: 69 Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 1426 ADR: 4.3 I Merck Index: 13,08664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...