Lithium metaborate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
205524-100G 205524-500G
SIAL205524-100GEA 162 EUR
SIAL205524-100G SIAL205524-500G
Lithium metaborate, Sigma-Aldrich®
Lithium metaborate
Formula: BLiO₂
MW: 49.75 g/mol
Boiling Pt: 1719
Melting Pt: 845
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011089
CAS Number: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546