Sodium tetraphenylborate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt
72020-250G 72020-50G
SIAL72020-250GEA 1130 EUR
SIAL72020-250G SIAL72020-50G
Sodium tetraphenylborate, Sigma-Aldrich®
Sodium tetraphenylborate
Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: > 400 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,08765