Sodium tetraphenylborate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt
M-Clarity™ quality level = MQ200
T25402-100G T25402-25G T25402-5G
SIALT25402-100GEA 309 EUR
SIALT25402-100G SIALT25402-25G SIALT25402-5G
Sodium tetraphenylborate, Sigma-Aldrich®
Sodium tetraphenylborate
Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: > 400 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,08765