Cesium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cesium Chloride
M-Clarity™ quality level = MQ200
C3032-100G C3032-1KG C3032-25G C3032-500G
SIALC3032-100GEA 222 EUR
SIALC3032-100G SIALC3032-1KG SIALC3032-25G SIALC3032-500G
Cesium chloride, Sigma-Aldrich®
Cesium chloride
Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
Density: 3.7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021