Potassium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Potassiumchloride
M-Clarity™ quality level = MQ100
449989-10G 449989-50G
SIAL449989-10GEA 164 EUR
SIAL449989-10G SIAL449989-50G
Potassium chloride, Sigma-Aldrich®
Potassium chloride
Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621