Potassium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Potassiumchloride
M-Clarity™ quality level = MQ100
451029-10G 451029-50G
SIAL451029-10GEA 77.3 EUR
SIAL451029-10G SIAL451029-50G
Potassium chloride, Sigma-Aldrich®
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621