Potassium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P5405-1KG P5405-250G P5405-500G
SIALP5405-1KGEA 143 EUR
SIALP5405-1KG SIALP5405-250G SIALP5405-500G
Potassium chloride, Sigma-Aldrich®
Potassium chloride
Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621