Potassium chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Potassiumchloride

M-Clarity™ quality level = MQ200

P9541-1KG P9541-500G P9541-5KG
SIALP9541-1KGEA 146 EUR
SIALP9541-1KG SIALP9541-500G SIALP9541-5KG
Potassium chloride, Sigma-Aldrich®
Potassium chloride
Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR