Sodium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Saline, NaCl, Sea salt, Halite
M-Clarity™ quality level = MQ200
S3014-10KG
S3014-1KG
S3014-500G
S3014-5KG
S3014-6X1KG
SIALS3014-10KGEA
729
EUR
SIALS3014-10KG
SIALS3014-1KG
SIALS3014-500G
SIALS3014-5KG
SIALS3014-6X1KG
Sodium chloride, Sigma-Aldrich®
Sodium chloride
Formula:
NaCl Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...