Sodium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
S6191-10KG S6191-1KG S6191-500G S6191-5KG
SIALS6191-10KGEA 908 EUR
SIALS6191-10KG SIALS6191-1KG SIALS6191-500G SIALS6191-5KG
Sodium chloride, Sigma-Aldrich®
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599