Potassium perchlorate 99
Supplier: Thermo Fisher Scientific
Danger
A11296.0E A11296.22 A11296.36
A11296.0EEA 314 EUR
A11296.0E A11296.22 A11296.36
Potassium perchlorate 99
Potassium perchlorate
Purity: 99% CAS No.: 7778-74-7 Molecular Formula: KClO4 Formula Weight: 138.55 Odour: Odourless Melting Point: 301-304° UN No.: 1489 MDL No.: MFCD00011362
Formula: ClKO₄
MW: 138.55 g/mol
Melting Pt: 301…304 °C
Density: 2.52
Storage Temperature: Ambient
MDL Number: MFCD00011362
CAS Number: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: 5.1 II
Merck Index: 13,07738