Cesium carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
554855-10G 554855-50G
SIAL554855-10GEA 74 EUR
SIAL554855-10G SIAL554855-50G
Cesium carbonate, Sigma-Aldrich®
Cesium carbonate
Formula: Cs₂CO₃
Melting Pt: 610 °C
Density: 4.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020