Sodium chromate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
307831-5G
307831-100G
307831-500G
SIAL307831-5GEA
34.3
EUR
SIAL307831-5G
SIAL307831-100G
SIAL307831-500G
Sodium chromate, Sigma-Aldrich®
Sodium chromate
Formula:
CrNa₂O₄ Melting Pt: 780 °C Density: 2.698 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003484 CAS Number: 7775-11-3 EINECS: 231-889-5 UN: 3288 ADR: 6.1,II Merck Index: 12,08745 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...