Potassium fluoride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
229814-100G 229814-10G
SIAL229814-100GEA 850 EUR
SIAL229814-100G SIAL229814-10G
Potassium fluoride, Sigma-Aldrich®
Potassium fluoride
Formula: KF
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714