Lithium fluoride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
203645-25G 203645-5G
SIAL203645-25GEA 222 EUR
SIAL203645-25G SIAL203645-5G
Lithium fluoride, Sigma-Aldrich®
Lithium fluoride
Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676
Melting Pt: 870
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,05553