Lithium fluoride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
449903-10G 449903-2G
SIAL449903-10GEA 244 EUR
SIAL449903-10G SIAL449903-2G
Lithium fluoride, Sigma-Aldrich®
Lithium fluoride
Formula: FLi
Boiling Pt: 1676 °C (1013 hPa)
Melting Pt: 870 °C
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,05553