Sodium fluoride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
S6776-100G S6776-500G
SIALS6776-100GEA 91.5 EUR
SIALS6776-100G SIALS6776-500G
Sodium fluoride, Sigma-Aldrich®
Sodium fluoride
Formula: NaF
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM