Cesium iodide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cesiumiodide
M-Clarity™ quality level = MQ100
203033-10G 203033-1G
SIAL203033-10GEA 124 EUR
SIAL203033-10G SIAL203033-1G
Cesium iodide, Sigma-Aldrich®
Cesium iodide
Formula: CsI
MW: 259.81 g/mol
Boiling Pt: 1280
Melting Pt: 621
Density: 3.21 g/cm³ (20 °C)
MDL Number: MFCD00010962
CAS Number: 7789-17-5
EINECS: 232-145-2
Merck Index: 12,02056