Lithium iodide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
439746-25G 439746-5G
SIAL439746-25GEA 555 EUR
SIAL439746-25G SIAL439746-5G
Lithium iodide, Sigma-Aldrich®
Lithium iodide
Formula: ILi
MW: 133.85 g/mol
Boiling Pt: 1177
Melting Pt: 450
Density: 3.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Merck Index: 13,05557