Sodium iodide

Supplier: Roth Carl
Warning

A134.1 A134.2
ROTHA134.1EA 236 EUR
ROTHA134.1 ROTHA134.2
Sodium iodide
Sodium iodide
Formula: NaI
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR