Sodium iodide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
M-Clarity™ quality level = MQ100
409286-100G
409286-10G
409286-1G
SIAL409286-100GEA
465
EUR
SIAL409286-100G
SIAL409286-10G
SIAL409286-1G
Sodium iodide, Sigma-Aldrich®
Sodium iodide
Formula:
NaI MW: 149.89 g/mol Boiling Pt: 1300 Melting Pt: 662 Density: 3.67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9 III Merck Index: 13,08705 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...