Trilithium nitride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
399558-10G 399558-25G 399558-5G
SIAL399558-10GEA 112 EUR
SIAL399558-10G SIAL399558-25G SIAL399558-5G
Trilithium nitride, Sigma-Aldrich®
tri-Lithium nitride
Formula: Li₃N
MW: 34.83 g/mol
Melting Pt: 845
Density: 1.38 g/cm³ (20 °C)
MDL Number: MFCD00016186
CAS Number: 26134-62-3
EINECS: 247-475-2
UN: 2806
ADR: 4.3 I