Sodium azide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
S8032-100G S8032-25G S8032-500G
SIALS8032-100GEA 160 EUR
SIALS8032-100G SIALS8032-25G SIALS8032-500G
Sodium azide, Sigma-Aldrich®
Sodium azide
Formula: NaN₃
Melting Pt: 275 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 13,08653