Sodium nitrite, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Diazotizing salts
M-Clarity™ quality level = MQ200
563218-100G 563218-25G
SIAL563218-100GEA 424 EUR
SIAL563218-100G SIAL563218-25G
Sodium nitrite, Sigma-Aldrich®
Sodium nitrite
Formula: NaNO₂
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 284 °C
Density: 2.168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721