, (dried basis) 98+%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
A11321.0B A11321.0I A11321.30
A11321.0BEA 146 EUR
A11321.0B A11321.0I A11321.30
, (dried basis) 98+%
di-Potassium hydrogen phosphate
Purity: 98+% CAS No.: 7758-11-4 Molecular Formula: K2HPO4 Formula Weight: 174.18 Merck Reference: 14,7658 MDL No.: MFCD00011383
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 14,7658
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM