Potassium tetrachloroplatinate(II), Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate
M-Clarity™ quality level = MQ100
520853-1G
520853-25G
520853-5G
SIAL520853-1GEA
165
EUR
SIAL520853-1G
SIAL520853-25G
SIAL520853-5G
Potassium tetrachloroplatinate(II), Sigma-Aldrich®
Potassium tetrachloroplatinate (II)
Formula:
Cl₄K₂Pt Boiling Pt: 35…40 °C (20 mmHg) Melting Pt: 265 °C Density: 3.38 g/cm³ (20 °C) |
MDL Number:
MFCD00011378 CAS Number: 10025-99-7 EINECS: 233-050-9 UN: 3288 ADR: 6.1,III Merck Index: 12,07855 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...