Potassium hexachloroplatinate(IV), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Dipotassium hexachloroplatinate
M-Clarity™ quality level = MQ100
206067-1G 206067-25G
SIAL206067-1GEA 119 EUR
SIAL206067-1G SIAL206067-25G
Potassium hexachloroplatinate(IV), Sigma-Aldrich®
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
MW: 485.99 g/mol
Melting Pt: 250
Density: 3.344 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3290
ADR: 6.1 II
Merck Index: 13,07720