Potassium hexachloroplatinate(IV), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Dipotassium hexachloroplatinate
M-Clarity™ quality level = MQ100
379859-1G 379859-5G
SIAL379859-1GEA 199 EUR
SIAL379859-1G SIAL379859-5G
Potassium hexachloroplatinate(IV), Sigma-Aldrich®
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
MW: 485.99 g/mol
Melting Pt: 250
Density: 3.344 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3290
ADR: 6.1 II
Merck Index: 13,07720