Potassium perrhenate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
243590-25G 243590-5G
SIAL243590-25GEA 787 EUR
SIAL243590-25G SIAL243590-5G
Potassium perrhenate, Sigma-Aldrich®
Potassium perrhenate
Formula: KO₄Re
Boiling Pt: 1370 °C (1013 hPa)
Melting Pt: 550 °C
Density: 4.88 g/cm³ (20 °C)
MDL Number: MFCD00011365
CAS Number: 10466-65-6
EINECS: 233-953-8
UN: 1479
ADR: 5.1,II