Sodium peroxodisulphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt
M-Clarity™ quality level = MQ100
S6172-1KG S6172-250G
SIALS6172-1KGEA 279 EUR
SIALS6172-1KG SIALS6172-250G
Sodium peroxodisulphate, Sigma-Aldrich®
Sodium peroxodisulphate
Formula: Na₂S₂O₈
MW: 238.11 g/mol
Melting Pt: 100
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003501
CAS Number: 7775-27-1
EINECS: 231-892-1
UN: 1505
ADR: 5.1 III
Merck Index: 13,08729