Potassium hydrogen sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Potassium bisulphate
401544-10G 401544-50G
SIAL401544-10GEA 48.4 EUR
SIAL401544-10G SIAL401544-50G
Potassium hydrogen sulphate, Sigma-Aldrich®
Potassium hydrogen sulphate
Formula: KHSO₄
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695