Sodium hydrogen sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium bisulphate
M-Clarity™ quality level = MQ200
307823-2.5KG 307823-20KG 307823-500G
SIAL307823-2.5KGEA 218 EUR
SIAL307823-2.5KG SIAL307823-20KG SIAL307823-500G
Sodium hydrogen sulphate, Sigma-Aldrich®
Sodium hydrogen sulphate
Formula: HNaO₄S
Melting Pt: 181.85 °C
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003531
CAS Number: 7681-38-1
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 12,08729