Magnesium diboride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Magnesium boride

M-Clarity™ quality level = MQ100

553913-25G 553913-5G
SIAL553913-25GEA 226 EUR
SIAL553913-25G SIAL553913-5G
Magnesium diboride, Sigma-Aldrich®
Magnesium diboride
Formula: B₂Mg
Density: 2.3 g/cm³ (20 °C)
MDL Number: MFCD00049752
CAS Number: 12007-25-9
EINECS: 234-501-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR