Barium bromide, anhydrous 99%, pure
Supplier: Thermo Fisher Scientific
Warning
315380500 315382500
ACRO315380500EA 77.8 EUR
ACRO315380500 ACRO315382500
Barium bromide, anhydrous 99%, pure
Barium bromide
Formula: BaBr₂
MW: 297.14 g/mol
Boiling Pt: 1835 °C (1013 hPa)
Melting Pt: 850 °C
Storage Temperature: Ambient
MDL Number: MFCD00003444
CAS Number: 10553-31-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,00995
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Argentometric ≥98.5 %