Strontium chloride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
439665-25G
439665-5G
SIAL439665-25GEA
389
EUR
SIAL439665-25G
SIAL439665-5G
Strontium chloride, Sigma-Aldrich®
Strontium chloride
Formula:
Cl₂Sr MW: 158.53 g/mol Boiling Pt: 1250 Melting Pt: 868 Density: 1.96 g/cm³ (20 °C) |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 EINECS: 233-971-6 Merck Index: 12,09000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...