Barium carbonate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
202711-100G
202711-25G
SIAL202711-100GEA
455
EUR
SIAL202711-100G
SIAL202711-25G
Barium carbonate, Sigma-Aldrich®
Barium carbonate
Formula:
BaCO₃ Melting Pt: 1350 °C Density: 3.89 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1,III Merck Index: 13,00972 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...