Barium carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
329436-25G 329436-100G
SIAL329436-25GEA 112 EUR
SIAL329436-25G SIAL329436-100G
Barium carbonate, Sigma-Aldrich®
Barium carbonate
Formula: BaCO₃
Melting Pt: 1350 °C
Density: 3.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972