Calcium carbonate, Sigma-Aldrich®
Supplier: Merck
21061-1KG-F 21061-250G-F 21061-50G-F
SIAL21061-1KG-FEA 423 EUR
SIAL21061-1KG-F SIAL21061-250G-F SIAL21061-50G-F
Calcium carbonate, Sigma-Aldrich®
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658