Strontium fluoride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
450030-10G 450030-50G
SIAL450030-10GEA 135 EUR
SIAL450030-10G SIAL450030-50G
Strontium fluoride, Sigma-Aldrich®
Formula: F₂Sr
Boiling Pt: 2460 °C (1013 hPa)
Melting Pt: 1477 °C
Density: 4.24 g/cm³ (20 °C)
MDL Number: MFCD00011251
CAS Number: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III
Merck Index: 12,09002