Calcium hydride ≥98% (metals basis excluding Mg) (max. 1% Mg)
Supplier: Thermo Fisher Scientific
|
Danger
|
41645.18
41645.30
41645.18EA
283
EUR
41645.18
41645.30
Calcium hydride ≥98% (metals basis excluding Mg) (max. 1% Mg)
Calcium hydride
Purity: 98%
CAS No.: 7789-78-8
Molecular Formula: CaH2
Formula Weight: 42.10
Physical Form: -10 Mesh
Density: 1.900
Melting Point: 600° dec.
Merck Reference: 14,1672
UN No.: 1404
MDL No.: MFCD00010897
Formula:
H₂Ca Melting Pt: 600 °C (dec.) Density: 1.900 Storage Temperature: Ambient |
MDL Number:
MFCD00010897 CAS Number: 7789-78-8 EINECS: 232-189-2 UN: 1404 ADR: 4.3,I Merck Index: 14,1672 |