Strontium iodide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
466336-10G 466336-2G
SIAL466336-10GEA 313 EUR
SIAL466336-10G SIAL466336-2G
Strontium iodide, Sigma-Aldrich®
Strontium iodide
Formula: I₂Sr
MW: 341.43 g/mol
Melting Pt: 515
Density: 4.437 g/cm³ (25 °C)
MDL Number: MFCD00049557
CAS Number: 10476-86-5
EINECS: 233-972-1
UN: 3260
ADR: 8 II
Merck Index: 12,09005