Barium nitrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
202754-100G 202754-25G
SIAL202754-100GEA 185 EUR
SIAL202754-100G SIAL202754-25G
Barium nitrate, Sigma-Aldrich®
Barium nitrate
Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: ∼592 °C
Density: ∼3.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 II
Merck Index: 13,00986