Calcium oxide, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

634182-100G 634182-25G
SIAL634182-100GEA 555 EUR
SIAL634182-100G SIAL634182-25G
Calcium oxide, Sigma-Aldrich®
Calcium oxide
Formula: CaO
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2580 °C
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR