Magnesium hydroxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
632309-100G 632309-25G
SIAL632309-100GEA 856 EUR
SIAL632309-100G SIAL632309-25G
Magnesium hydroxide, Sigma-Aldrich®
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.3261…2.3631 g/cm³ (20 °C)
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693