Magnesium sulphate, anhydrous ≥98%
Supplier: MP Biomedicals
Synonyms: MgSO4, Magnesium sulphate anhydrous
0215013691 0215013690
ICNA0215013691EA 119 EUR
ICNA0215013691 ICNA0215013690
Magnesium sulphate, anhydrous ≥98%
Magnesium sulphate
Soluble in water.
Formula: MgSO₄
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM