Magnesium sulphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: MgSO4, Magnesium sulphate anhydrous
M-Clarity™ quality level = MQ100
203726-125G 203726-25G
SIAL203726-125GEA 1010 EUR
SIAL203726-125G SIAL203726-25G
Magnesium sulphate, Sigma-Aldrich®
Magnesium sulphate
Formula: MgSO₄
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM