Aluminium oxide, nanopowder <50 nm (TEM), Sigma-Aldrich®
Supplier: Merck
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
M-Clarity™ quality level = MQ200
544833-10G 544833-50G
SIAL544833-10GEA 35.7 EUR
SIAL544833-10G SIAL544833-50G
Aluminium oxide, nanopowder <50 nm (TEM), Sigma-Aldrich®
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355