Aluminium oxide ≥99.99%, single crystal substrate <0001>, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
M-Clarity™ quality level = MQ100
634875-1EA 634875-5EA
SIAL634875-1EAEA 149 EUR
SIAL634875-1EA SIAL634875-5EA
Aluminium oxide ≥99.99%, single crystal substrate <0001>, Sigma-Aldrich®
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355