Lead(II) bromide, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

211141-100G 211141-500G
SIAL211141-100GEA 41.3 EUR
SIAL211141-100G SIAL211141-500G
Lead(II) bromide, Sigma-Aldrich®
Lead (II) bromide
Formula: Br₂Pb
Boiling Pt: 916 °C (1013 hPa)
Melting Pt: 362 °C
Density: 6.676 g/cm³ (0 °C)
MDL Number: MFCD00011156
CAS Number: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1,III
Merck Index: 12,05418

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR